1,4-Bis(methoxymethyl)benzene ≥99.0% (by GC)
Fornitore: TCI
Formula:
C₁₀H₁₄O₂ Punto di ebollizione: 235°C Flash Pt: 118°C |
MDL Number:
MFCD00025903 CAS Number: 6770-38-3 EINECS: 229-828-2 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...