Stampa…

Bis(1,3-propanediamine)copper(II) dichloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

B1513-1G B1513-10G
TCIAB1513-1GEA 29 EUR
TCIAB1513-1G TCIAB1513-10G
Bis(1,3-propanediamine)copper(II) dichloride ≥98.0% (by titrimetric analysis)
Bis(1,3-propanediamine)copper(II) dichloride
Formula: C₆H₂₀Cl₂CuN₄
Peso molecolare: 282,70 g/mol
MDL Number: MFCD00191318
CAS Number: 32270-93-2

Visualizza i codici

Risultati del test di specificazione

Appearance Blue to Purple powder to crystal
Purity (Chelometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR