Stampa…

1,3-Bis(4,5-dihydro-2-oxazolyl)benzene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,2'-(1,3-Phenylene)bis(2-oxazoline), 1,3-Bis(4,5-dihydrooxazol-2-yl)benzene

B1511-25G B1511-500G B1511-100G
TCIAB1511-25GEA 17 EUR
TCIAB1511-25G TCIAB1511-500G TCIAB1511-100G
1,3-Bis(4,5-dihydro-2-oxazolyl)benzene ≥98.0% (by GC)
1,3-Bis(4,5-dihydro-2-oxazolyl)benzene
Formula: C₁₂H₁₂N₂O₂
Peso molecolare: 216,24 g/mol
MDL Number: MFCD00191606
CAS Number: 34052-90-9
EINECS: 421-510-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 146.0 to 149.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR