Stampa…

Ethyl-tert-butylacetate ≥99.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: tert-Butylacetic Acid Ethyl Ester, Ethyltert-butylacetate, 3,3-Dimethylbutyric Acid Ethyl Ester, Ethyl 3,3-Dimethylbutyrate

B1510-100ML B1510-25ML
TCIAB1510-100MLEA 248 EUR
TCIAB1510-100ML TCIAB1510-25ML
Ethyl-tert-butylacetate ≥99.0% (by GC)
Ethyl-tert-butylacetate
Formula: C₈H₁₆O₂
Peso molecolare: 144,21 g/mol
Punto di ebollizione: 145°C
Flash Pt: 34°C
MDL Number: MFCD00026911
CAS Number: 5340-78-3
EINECS: 226-280-6
UN: 3272
ADR: 3,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR