Stampa…

1,3-Bis(4-hydroxyphenoxy)benzene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Resorcinol Bis(4-hydroxyphenyl) Ether

B1484-1G B1484-5G
TCIAB1484-1GEA 129 EUR
TCIAB1484-1G TCIAB1484-5G
1,3-Bis(4-hydroxyphenoxy)benzene ≥98.0% (by GC)
1,3-Bis(4-hydroxyphenoxy)benzene
Formula: C₁₈H₁₄O₄
Peso molecolare: 294,31 g/mol
MDL Number: MFCD00142505
CAS Number: 126716-90-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 135.0 to 139.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR