Stampa…

O-Benzyl-L-serine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: H-Ser(Bzl)-OMe.HCl, H-Ser(Bzl)-OMe·HCl, Methyl O-Benzyl-L-serinate Hydrochloride

B1450-1G B1450-5G
TCIAB1450-1GEA 65 EUR
TCIAB1450-1G TCIAB1450-5G
O-Benzyl-L-serine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
O-Benzyl-L-serine methyl ester hydrochloride
Formula: C₁₁H₁₅NO₃
Peso molecolare: 245,70 g/mol
MDL Number: MFCD00191312
CAS Number: 19525-87-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +4.0 to +6.0 deg (C=1, MeOH)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR