Stampa…

N-Benzyliminodiacetic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: 2,2'-(Benzylazanediyl)diacetic acid, N-Benzyliminodiacetic acid

B1363-25G B1363-5G
TCIAB1363-25GEA 271 EUR
TCIAB1363-25G TCIAB1363-5G
N-Benzyliminodiacetic acid ≥98.0% (by titrimetric analysis)
N-Benzyliminodiacetic acid
Formula: C₁₁H₁₃NO₄
Peso molecolare: 223.23 g/mol
MDL Number: MFCD00004285
CAS Number: 3987-53-9
EINECS: 223-631-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1347f5226d4a488b26abac5f0ffedae0cdb75af7&vid=1347f5226d4a488b26abac5f0ffedae0cdb75af7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point (Decomposition) 198.0 to 203.0 deg-C
Solubility in hot Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR