1-Benzyl-3-aminopyrrolidine ≥97.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₁₁H₁₆N₂ |
MDL Number:
MFCD00059036 CAS Number: 18471-40-4 UN: 2735 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...