Stampa…

3-Benzyloxazolidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

B1339-1G B1339-5G
TCIAB1339-1GEA 55 EUR
TCIAB1339-1G TCIAB1339-5G
3-Benzyloxazolidine ≥98.0% (by GC, titration analysis)
3-Benzyloxazolidine
Formula: C₁₀H₁₃NO
Peso molecolare: 163,22 g/mol
Punto di ebollizione: 127°C
MDL Number: MFCD00059028
CAS Number: 13657-16-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR