4-tert-Butylphthalonitrile ≥98.0% (by GC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-(tert-Butyl)phthalonitrile, 4-tert-Butyl-1,2-dicyanobenzene
B1334-25G
B1334-5G
TCIAB1334-25GEA
368
EUR
TCIAB1334-25G
TCIAB1334-5G
4-tert-Butylphthalonitrile ≥98.0% (by GC, total nitrogen)
4-tert-Butylphthalonitrile
Formula:
C₁₂H₁₂N₂ Peso molecolare: 184,24 g/mol Punto di ebollizione: 165°C |
MDL Number:
MFCD00060112 CAS Number: 32703-80-3 EINECS: 251-166-8 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...