Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
B1317-25G
B1317-500G
TCIAB1317-25GEA
36
EUR
TCIAB1317-25G
TCIAB1317-500G
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride ≥98.0% (by titrimetric analysis)
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride
Formula:
C₁₂H₈O₆ Peso molecolare: 248,19 g/mol |
MDL Number:
MFCD00082228 CAS Number: 1719-83-1 EINECS: 217-009-2 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Methanolysis Method) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...