Stampa…

Bis(ethylenedithio)tetrathiafulvalene for organic electronics

Fornitore: TCI EUROPE

Synonyms: BEDT-TTF, Bis(ethylenedithiolo)tetrathiafulvalene

B1200-1G B1200-100MG B1200-5G
TCIAB1200-1GEA 149 EUR
TCIAB1200-1G TCIAB1200-100MG TCIAB1200-5G
Bis(ethylenedithio)tetrathiafulvalene for organic electronics
Bis(ethylenedithio)tetrathiafulvalene
Formula: C₁₀H₈S₈
Peso molecolare: 384,65 g/mol
MDL Number: MFCD00059710
CAS Number: 66946-48-3

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Amber to Dark red powder to crystal
Elemental analysis (Carbon) 30.00 to 32.00 %
Elemental analysis (Sulfuride) 65.00 to 69.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR