4,5-Bis(benzoylthio)-1,3-dithiole-2-thione ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
B1199-1G
B1199-5G
TCIAB1199-1GEA
71
EUR
TCIAB1199-1G
TCIAB1199-5G
4,5-Bis(benzoylthio)-1,3-dithiole-2-thione ≥98.0% (by HPLC, titration analysis)
4,5-Bis(benzoylthio)-1,3-dithiole-2-thione
Formula:
C₁₇H₁₀O₂S₅ Peso molecolare: 406,57 g/mol |
MDL Number:
MFCD00059894 CAS Number: 68494-08-6 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 140.0 to 143.0 deg-C |
Solubility in Chloroform | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...