Stampa…

Boc-L-isoleucine, hemihydrate ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: N-Boc-L-isoleucine, Boc-Ile-OH

B1186-25G B1186-5G
TCIAB1186-25GEA 52 EUR
TCIAB1186-25G TCIAB1186-5G
Boc-L-isoleucine, hemihydrate ≥99.0% (by titrimetric analysis)
Boc-L-isoleucine
Formula: C₁₁H₂₁NO₄·½H₂O
Peso molecolare: 231,06 g/mol
MDL Number: MFCD00038324
CAS Number: 13139-16-7
EINECS: 236-074-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0% (calcd.on anh.substance)
Melting point 59.0 to 63.0 deg-C
Specific rotation [a]20/D +2.4 to +3.0 deg (C=2, AcOH)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR