Stampa…

N-Boc-L-alanine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: N-Alpha-t-Boc-L-alanine, (S)-2-((tert-Butoxycarbonyl)amino)propanoic acid, BOC-L-Alanine, Boc-Ala-OH

B1184-25G B1184-5G
TCIAB1184-25GEA 39 EUR
TCIAB1184-25G TCIAB1184-5G
N-Boc-L-alanine ≥98.0% (by HPLC, titration analysis)
N-Boc-L-alanine
Formula: C₈H₁₅NO₄
Peso molecolare: 189,21 g/mol
MDL Number: MFCD00037225
CAS Number: 15761-38-3
EINECS: 239-847-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 81.0 to 85.0 deg-C
Specific rotation [a]20/D -24.0 to -27.0 deg (C=2, AcOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR