Stampa…
N-[4-(2-Benzimidazolyl)phenyl]maleimide ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Attenzione
B1149-100MG B1149-50MG
TCIAB1149-100MGEA 200 EUR
TCIAB1149-100MG TCIAB1149-50MG
N-[4-(2-Benzimidazolyl)phenyl]maleimide ≥98.0% (by HPLC, titration analysis)
N-[4-(2-Benzimidazolyl)phenyl]maleimide
Formula: C₁₇H₁₁N₃O₂
Peso molecolare: 289,29 g/mol
Punto di fusione: 247°C
MDL Number: MFCD00042767
CAS Number: 27030-97-3
EINECS: 248-173-3
Visualizza i codici
Appearance Light yellow to Amber powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %