Benzoin isobutyl ether ≥94.0% (by GC)
Fornitore: TCI
Formula:
C₁₈H₂₀O₂ |
MDL Number:
MFCD00008933 CAS Number: 22499-12-3 EINECS: 245-039-6 |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow to Light orange powder to lump to clear liquid |
Purity (GC) | min. 94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...