Stampa…

1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Hexahydro-m-xylylenediamine, 1,3-Di(aminomethyl)cyclohexane, Cyclohexane-1,3-diyldimethanamine

B1005-25ML B1005-500ML
TCIAB1005-25MLEA 17 EUR
TCIAB1005-25ML TCIAB1005-500ML
1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture)
Formula: C₈H₁₈N₂
Peso molecolare: 142,25 g/mol
Flash Pt: 113°C
MDL Number: MFCD00001522
CAS Number: 2579-20-6
EINECS: 219-941-5
UN: 2735
ADR: 8,I

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR