O-Benzyl-L-serine ≥99.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
H-Ser(Bzl)-OH
B0861-1G
B0861-5G
TCIAB0861-1GEA
22
EUR
TCIAB0861-1G
TCIAB0861-5G
O-Benzyl-L-serine ≥99.0% (by titrimetric analysis)
O-Benzyl-L-serine
Formula:
C₁₀H₁₃NO₃ Peso molecolare: 195,22 g/mol |
MDL Number:
MFCD00065937 CAS Number: 4726-96-9 EINECS: 225-220-6 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Specific rotation [a]20/D | +6.5 to +8.0 deg (C=2, 1mol/L HCl) |
Solubility in hot Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...