Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
Bz-Arg-OEt.HCl, Nα-Benzoyl-L-arginine Ethyl Ester HCl
B0853-25G
B0853-5G
TCIAB0853-25GEA
106
EUR
TCIAB0853-25G
TCIAB0853-5G
Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥98.0% (by titrimetric analysis)
Nalpha-Benzoyl-L-arginine ethyl ester hydrochloride
Formula:
C₁₅H₂₂N₄O₃ Peso molecolare: 342,82 g/mol |
MDL Number:
MFCD00012579 CAS Number: 2645-08-1 EINECS: 220-157-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Specific rotation [a]20/D | -14.0 to -20.0 deg (C=2, H2O) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...