Stampa…

Azoic diazo component 24 (salt) ≥95.0% (by titrimetric analysis) for biochemical research

Fornitore: TCI

B0785-25G B0785-5G
TCIAB0785-25GEA 105 EUR
TCIAB0785-25G TCIAB0785-5G
Azoic diazo component 24 (salt) ≥95.0% (by titrimetric analysis) for biochemical research
Azoic diazo component 24 (salt)
Formula: C₁₅H₁₄ClN₃O₃
MDL Number: MFCD00074762
CAS Number: 55663-99-5

Visualizza i codici

Risultati del test di specificazione

Appearance Yellow to Brown to Dark green powder to crystal
Purity (Argentmetric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR