Vinyl butyrate ≥98.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Butyric Acid Vinyl Ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
B0765-25ML
B0765-500ML
B0765-100ML
TCIAB0765-25MLEA
21
EUR
TCIAB0765-25ML
TCIAB0765-500ML
TCIAB0765-100ML
Vinyl butyrate ≥98.0% (by GC) stabilizzato
Vinyl butyrate
Formula:
C₆H₁₀O₂ Peso molecolare: 114,14 g/mol Punto di ebollizione: 116°C Flash Pt: 20°C |
MDL Number:
MFCD00059410 CAS Number: 123-20-6 EINECS: 204-609-4 UN: 2838 ADR: 3,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...