Stampa…

Butyl phthalyl butyl glycolate ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Phthalic Acid Butoxycarbonylmethyl Butyl Ester, Butoxycarbonylmethyl Butyl Phthalate

B0737-25G B0737-500G
TCIAB0737-25GEA 18 EUR
TCIAB0737-25G TCIAB0737-500G
Butyl phthalyl butyl glycolate ≥95.0% (by GC)
Butyl phthalyl butyl glycolate
Formula: C₁₈H₂₄O₆
Peso molecolare: 336,38 g/mol
Punto di ebollizione: 219°C
Flash Pt: 199°C
MDL Number: MFCD00053793
CAS Number: 85-70-1
EINECS: 201-624-8

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %
Acid Value max. 0.1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR