Stampa…

2,4-Diamino-6-butylamino-1,3,5-triazine ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N2-Butyl-1,3,5-triazine-2,4,6-triamine, N2-Butylmelamine, 2-Butylamino-4,6-diamino-1,3,5-triazine

B0727-25G B0727-5G
TCIAB0727-25GEA 224 EUR
TCIAB0727-25G TCIAB0727-5G
2,4-Diamino-6-butylamino-1,3,5-triazine ≥97.0% (by titrimetric analysis)
2,4-Diamino-6-butylamino-1,3,5-triazine
Formula: C₇H₁₄N₆
Peso molecolare: 182,23 g/mol
MDL Number: MFCD00059764
CAS Number: 5606-24-6
EINECS: 227-025-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 93.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR