Stampa…

2-Bromohexanoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: DL-2-Bromohexanoic acid

B0601-25G B0601-500G
TCIAB0601-25GEA 22 EUR
TCIAB0601-25G TCIAB0601-500G
2-Bromohexanoic acid ≥98.0% (by GC, titration analysis)
2-Bromohexanoic acid
Formula: C₆H₁₁BrO₂
Peso molecolare: 195,06 g/mol
MDL Number: MFCD00004218
CAS Number: 616-05-7
EINECS: 210-461-1
UN: 3265
ADR: 8,III
Merck Index: 14,01415

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR