Stampa…

Bindschedler's green leuco base ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Bis(4-dimethylaminophenyl)amine, Leuco Bindschedler's Green, N,N,N',N'-Tetramethyldiaminodiphenylamine

B0482-1G B0482-5G
TCIAB0482-1GEA 168 EUR
TCIAB0482-1G TCIAB0482-5G
Bindschedler's green leuco base ≥98.0% (by HPLC, titration analysis)
Bindschedler's green leuco base
Formula: C₁₆H₂₁N₃
Peso molecolare: 255,37 g/mol
MDL Number: MFCD00059291
CAS Number: 637-31-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 119.0 to 123.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR