Bis(cyclohexanone)oxalyldihydrazone ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Cupferazone, N'1,N'2-Dicyclohexylideneoxalohydrazide, Oxalic acid bis(cyclohexylidenehydrazide), Bis(cyclohexanone)oxaldihydrazone, Cuprizone
B0476-1G
B0476-25G
TCIAB0476-1GEA
25
EUR
TCIAB0476-1G
TCIAB0476-25G
Bis(cyclohexanone)oxalyldihydrazone ≥98.0% (by titrimetric analysis)
Bis(cyclohexanone)oxalyldihydrazone
Formula:
C₁₄H₂₂N₄O₂ Peso molecolare: 278,36 g/mol |
MDL Number:
MFCD00001659 CAS Number: 370-81-0 EINECS: 206-729-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...