Stampa…

3,3'-Oxydipropionitrile ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: Bis(2-cyanoethyl) Ether

B0475-25ML B0475-500ML
TCIAB0475-25MLEA 87 EUR
TCIAB0475-25ML TCIAB0475-500ML
3,3'-Oxydipropionitrile ≥98.0% (by GC)
3,3'-Oxydipropionitrile
Formula: C₆H₈N₂O
MDL Number: MFCD00001960
CAS Number: 1656-48-0
EINECS: 216-750-9

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR