Berberine chloride hydrate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
Natural Yellow 18
B0450-25G
B0450-5G
TCIAB0450-25GEA
51
EUR
TCIAB0450-25G
TCIAB0450-5G
Berberine chloride hydrate ≥98.0% (by HPLC, titration analysis)
Berberine chloride hydrate
Formula:
C₂₀H₁₈ClNO₄ Peso molecolare: 371,82 g/mol |
MDL Number:
MFCD00011939 CAS Number: 141433-60-5 Merck Index: 14,01154 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Solubility in hot Water | almost transparency |
Water | 8.0 to 12.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...