Stampa…

2-(Benzyloxy)ethanol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Ethylene Glycol Monobenzyl Ether, Benzyl Cellosolve, Benzyl Glycol

B0426-25ML B0426-250ML
TCIAB0426-25MLEA 21 EUR
TCIAB0426-25ML TCIAB0426-250ML
2-(Benzyloxy)ethanol ≥98.0% (by GC)
2-(Benzyloxy)ethanol
Formula: C₉H₁₂O₂
Peso molecolare: 152,19 g/mol
Punto di ebollizione: 256°C
Flash Pt: 130°C
MDL Number: MFCD00002868
CAS Number: 622-08-2
EINECS: 210-719-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26467f4aeb1d68f57d63ebdc026a7bf98b695cb6&vid=26467f4aeb1d68f57d63ebdc026a7bf98b695cb6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR