Stampa…

N-Benzoyl-DL-phenylalanine-2-naphthyl ester ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: N-Benzoyl-DL-phenylalanine β-naphthyl ester

B0309-1G B0309-100MG
TCIAB0309-1GEA 182 EUR
TCIAB0309-1G TCIAB0309-100MG
N-Benzoyl-DL-phenylalanine-2-naphthyl ester ≥98.0% (by HPLC)
N-Benzoyl-DL-phenylalanine-2-naphthyl ester
Formula: C₂₆H₂₁NO₃
Peso molecolare: 395,46 g/mol
MDL Number: MFCD00021607
CAS Number: 15873-25-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 156.0 to 159.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR