Stampa…

3,5-Diphenyl-1-(4-biphenylyl)formazan ≥95.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 3,5-Diphenyl-1-(p-diphenylyl)formazan

B0277-1G B0277-100MG
TCIAB0277-1GEA 175 EUR
TCIAB0277-1G TCIAB0277-100MG
3,5-Diphenyl-1-(4-biphenylyl)formazan ≥95.0% (by HPLC)
3,5-Diphenyl-1-(4-biphenylyl)formazan
Formula: C₂₅H₂₀N₄
Peso molecolare: 376,46 g/mol
MDL Number: MFCD00059680
CAS Number: 21520-85-4

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Brown to Dark purple powder to crystal
Purity (HPLC) min. 95.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR