Stampa…

Glycine benzyl ester-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Benzyl glycinate 4-methylbenzenesulphonate, H-Gly-OBzl.Tos-OH, H-Gly-OBzl.TosOH

B0266-25G B0266-5G
TCIAB0266-25GEA 70 EUR
TCIAB0266-25G TCIAB0266-5G
Glycine benzyl ester-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)
Glycine benzyl ester-p-toluenesulphonate
Formula: C₉H₁₁NO₂
Peso molecolare: 337,39 g/mol
MDL Number: MFCD00035425
CAS Number: 1738-76-7
EINECS: 217-094-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR