Benzyl butyl ether ≥95.0% (by GC)
Fornitore: TCI
Formula:
C₁₁H₁₆O Punto di ebollizione: 220°C Flash Pt: 91°C |
MDL Number:
MFCD00039954 CAS Number: 588-67-0 EINECS: 209-626-0 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...