Stampa…

Ethyl benzenesulphonate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Benzenesulphonic Acid Ethyl Ester

B0032-25G B0032-500G B0032-100G
TCIAB0032-25GEA 29 EUR
TCIAB0032-25G TCIAB0032-500G TCIAB0032-100G
Ethyl benzenesulphonate ≥98.0% (by GC)
Ethyl benzenesulphonate
Formula: C₈H₁₀O₃S
Peso molecolare: 186,23 g/mol
Punto di ebollizione: 156°C
MDL Number: MFCD00025033
CAS Number: 515-46-8
EINECS: 208-200-1
Merck Index: 14,01070

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Ester Value) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR