Stampa…

Sodium-9,10-dimethoxyanthracene-2-sulphonate ≥98.0% (by HPLC) fluorimetric ion-pair reagent for amines

Fornitore: TCI

Synonyms: IPA-DAS, 9,10-Dimethoxyanthracene-2-sulphonic acid Sodium Salt

A5701-1G A5701-5G A5701-200MG
TCIAA5701-1GEA 156 EUR
TCIAA5701-1G TCIAA5701-5G TCIAA5701-200MG
Sodium-9,10-dimethoxyanthracene-2-sulphonate ≥98.0% (by HPLC) fluorimetric ion-pair reagent for amines
Sodium-9,10-dimethoxyanthracene-2-sulphonate
Formula: C₁₆H₁₄O₅S
MDL Number: MFCD00042061
CAS Number: 67580-39-6
EINECS: 266-720-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09f11b5dabf18afb7cc9b423cb6613803f21f3cf&vid=09f11b5dabf18afb7cc9b423cb6613803f21f3cf&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 95.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR