Stampa…

(S)-(+)-DBD-Py-NCS [=(S)-(+)-4-(N,N-Dimethylaminosulphonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)-2,1,3-benzoxadiazole] ≥98.0% (by HPLC) for HPLC labeling

Fornitore: TCI EUROPE

Synonyms: (S)-(+)-4-(N,N-Dimethylaminosulphonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)-2,1,3-benzoxadiazole, (S)-(+)-4-(N,N-Dimethylaminosulphonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)benzofurazan

A5569-100MG A5569-500MG
TCIAA5569-100MGEA 231 EUR
TCIAA5569-100MG TCIAA5569-500MG
(S)-(+)-DBD-Py-NCS [=(S)-(+)-4-(N,N-Dimethylaminosulphonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)-2,1,3-benzoxadiazole] ≥98.0% (by HPLC) for HPLC labeling
(S)-(+)-DBD-Py-NCS [=(S)-(+)-4-(N,N-Dimethylaminosulphonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)-2,1,3-benzoxadiazole]
Formula: C₁₃H₁₅N₅O₃S₂
Peso molecolare: 353,42 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00671494
CAS Number: 163927-32-0

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 157.0 to 161.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR