Stampa…

N-Chloromethyl-4-nitrophthalimide ≥98.0% (by titrimetric analysis) for HPLC labeling

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Nitro-N-chloromethylphthalimide

A5503-1G A5503-5G
TCIAA5503-1GEA 73 EUR
TCIAA5503-1G TCIAA5503-5G
N-Chloromethyl-4-nitrophthalimide ≥98.0% (by titrimetric analysis) for HPLC labeling
N-Chloromethyl-4-nitrophthalimide
Formula: C₉H₅ClN₂O₄
Peso molecolare: 240,60 g/mol
MDL Number: MFCD00059889
CAS Number: 54455-37-7

Visualizza i codici

Risultati del test di specificazione

Purity (Argentmetric Titration) min. 98.0 %
Melting point 153.0 to 157.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR