Fluoral-P (=4-amino-3-penten-2-one) ≥98.0% (by GC, titration analysis) fluorimetric reagent for aldehydes
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Amino-3-penten-2-one, 4-Aminopent-3-en-2-one
A5350-1G
A5350-5G
TCIAA5350-1GEA
20
EUR
TCIAA5350-1G
TCIAA5350-5G
Fluoral-P (=4-amino-3-penten-2-one) ≥98.0% (by GC, titration analysis) fluorimetric reagent for aldehydes
Fluoral-P (=4-amino-3-penten-2-one)
Formula:
C₅H₉NO Peso molecolare: 99,13 g/mol Punto di ebollizione: 214°C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00043715 CAS Number: 1118-66-7 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 40.0 to 42.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...