Stampa…

N-(4-Aminobutyl)-N-ethylisoluminol ≥97.0% (by titrimetric analysis) chemiluminescence reagent

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-[N-(4-Aminobutyl)-N-ethylamino]-2,3-dihydro-1,4-phthalazinedione, ABEI

A5304-1G A5304-100MG
TCIAA5304-1GEA 520 EUR
TCIAA5304-1G TCIAA5304-100MG
N-(4-Aminobutyl)-N-ethylisoluminol ≥97.0% (by titrimetric analysis) chemiluminescence reagent
N-(4-Aminobutyl)-N-ethylisoluminol
Formula: C₁₄H₂₀N₄O₂
Peso molecolare: 276,34 g/mol
MDL Number: MFCD00010559
CAS Number: 66612-29-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Sensitiveness Emits fluorescence in the presence of hydrogen peroxide and hemin

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR