Stampa…

8-Mercaptoquinoline hydrochloride ≥95.0% (by titrimetric analysis) per spettrofotometria, for fluorimetry

Fornitore: TCI

A5004-1G A5004-5G
TCIAA5004-1GEA 92 EUR
TCIAA5004-1G TCIAA5004-5G
8-Mercaptoquinoline hydrochloride ≥95.0% (by titrimetric analysis) per spettrofotometria, for fluorimetry
8-Mercaptoquinoline hydrochloride

Extraction-spectrophotometric and fluorimetric reagent for soft metals

Formula: C₉H₇NS
MDL Number: MFCD00043261
CAS Number: 34006-16-1
EINECS: 251-785-3

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (Neutralization titration) min. 95.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR