5-Adamantyl-IAA ≥98.0% (by HPLC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₂₀H₂₃NO₂ Temperatura di stoccaggio: Refrigerata |
CAS Number:
2244426-40-0 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...