Stampa…

2-(2-Amino-4-nitroanilino)ethanol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Amino-4-nitro-N-(2-hydroxyethyl)aniline, HC Yellow No. 5

A3332-25G A3332-5G
TCIAA3332-25GEA 68 EUR
TCIAA3332-25G TCIAA3332-5G
2-(2-Amino-4-nitroanilino)ethanol ≥98.0% (by GC, titration analysis)
2-(2-Amino-4-nitroanilino)ethanol
Formula: C₈H₁₁N₃O₃
Peso molecolare: 197,19 g/mol
MDL Number: MFCD00156329
CAS Number: 56932-44-6
EINECS: 260-450-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 133.0 to 137.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR