2-Amino-5-bromobenzophenone ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₃H₁₀BrNO |
MDL Number:
MFCD00059962 CAS Number: 39859-36-4 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 111.0 to 115.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...