Stampa…

Acetamidine hydrobromide ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Acetamidinium bromide

A3292-1G A3292-5G
TCIAA3292-1GEA 39 EUR
TCIAA3292-1G TCIAA3292-5G
Acetamidine hydrobromide ≥98.0% (by titrimetric analysis)
Acetamidine hydrobromide
Formula: C₂H₆N₂·HBr
Peso molecolare: 139,00 g/mol
CAS Number: 1040352-82-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR