Stampa…

3-Amino-2-chloro-6-methylphenol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Chloro-4-methyl-3-hydroxyaniline, 5-Amino-6-chloro-o-cresol

A3266-5G A3266-25G
TCIAA3266-5GEA 36 EUR
TCIAA3266-5G TCIAA3266-25G
3-Amino-2-chloro-6-methylphenol ≥98.0% (by GC, titration analysis)
3-Amino-2-chloro-6-methylphenol
Formula: C₇H₈ClNO
Peso molecolare: 157,60 g/mol
MDL Number: MFCD03094650
CAS Number: 84540-50-1
EINECS: 283-144-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR