Stampa…

2-Acetyl-2-thiazoline ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 1-(4,5-Dihydrothiazol-2-yl)ethanone

A3264-1G A3264-5G
TCIAA3264-1GEA 21 EUR
TCIAA3264-1G TCIAA3264-5G
2-Acetyl-2-thiazoline ≥98.0% (by GC, titration analysis)
2-Acetyl-2-thiazoline
Formula: C₅H₇NOS
Peso molecolare: 129,18 g/mol
Punto di ebollizione: 94°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00209496
CAS Number: 29926-41-8

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR