(S)-Methyl-2-acetamidopropanoate ≥98.0% (by GC)
Fornitore: TCI
A3262-5G
A3262-25G
TCIAA3262-5GEA
26
EUR
TCIAA3262-5G
TCIAA3262-25G
(S)-Methyl-2-acetamidopropanoate ≥98.0% (by GC)
(S)-Methyl-2-acetamidopropanoate
Formula:
C₆H₁₁NO₃ Punto di ebollizione: 90°C |
MDL Number:
MFCD00038239 CAS Number: 3619-02-1 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -98.0 to -88.0 deg (C=1,H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...