Stampa…

2-Aminopyridine-4-methanol ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-Amino-4-pyridinylmethanol, (2-Aminopyridin-4-yl)-methanol, (2-Aminopyridin-4-yl)methanol, (2-Aminopyrid-4-yl)methanol

A3249-1G A3249-5G
TCIAA3249-1GEA 35 EUR
TCIAA3249-1G TCIAA3249-5G
2-Aminopyridine-4-methanol ≥98.0% (by GC, titration analysis)
2-Aminopyridine-4-methanol
Formula: C₆H₈N₂O
MDL Number: MFCD03791261
CAS Number: 105250-17-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR