Stampa…

2-Amino-4-phenylpyrimidine ≥97.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 4-Phenylpyrimidin-2-amine

A3244-1G A3244-5G
TCIAA3244-1GEA 46 EUR
TCIAA3244-1G TCIAA3244-5G
2-Amino-4-phenylpyrimidine ≥97.0% (by GC, titration analysis)
2-Amino-4-phenylpyrimidine
Formula: C₁₀H₉N₃
MDL Number: MFCD00094706
CAS Number: 2305-87-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 162.0 to 166.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR